PRIH93ED84D8 Princeton BioMolecular Research, Inc.

N-(3-benzoylphenyl)-5-chloro-3-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonamide

  • MDL number MFCD07637999
  • Molecular Weight : 442.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart