PRIH93ED84DA Princeton BioMolecular Research, Inc.

N-(3-benzyl-4-oxo-3,4-dihydroquinazolin-6-yl)-3-fluoro-4-methoxybenzenesulfonamide

  • MDL number MFCD08287012
  • Molecular Weight : 439.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart