PRIH93ED84DC Princeton BioMolecular Research, Inc.

N-(3-benzoylphenyl)-4-methyl-3-nitrobenzenesulfonamide

  • MDL number MFCD12972267
  • Molecular Weight : 396.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart