PRIH93ED84DD Princeton BioMolecular Research, Inc.

N-[2-(2-chlorophenoxy)ethyl]naphthalene-2-sulfonamide

  • MDL number MFCD01025321
  • Molecular Weight : 361.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart