PRIH93ED84E1 Princeton BioMolecular Research, Inc.

3-(2-chlorophenyl)-2-[4-(dimethylamino)-3-nitrophenyl]-2,3-dihydroquinazolin-4(1H)-one

  • MDL number MFCD02615321
  • Molecular Weight : 422.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart