PRIH93ED84EC Princeton BioMolecular Research, Inc.

N-[4-(2-amino-2-oxoethoxy)phenyl]-3,4,5-trimethoxybenzamide

  • MDL number MFCD02625612
  • Molecular Weight : 360.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart