PRIH93ED84F0 Princeton BioMolecular Research, Inc.

(2E)-N-(4-chlorophenyl)-2-{[4-(diethylamino)phenyl]hydrazono}-3-oxobutanamide

  • MDL number MFCD02625655
  • Molecular Weight : 386.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart