PRIH93ED84F5 Princeton BioMolecular Research, Inc.

4-tert-butyl-N-(2-chloroethyl)benzenesulfonamide

  • MDL number MFCD02625668
  • Molecular Weight : 275.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart