PRIH93ED8502 Princeton BioMolecular Research, Inc.

4-chloro-N-(2-chloroethyl)-2,5-dimethoxybenzenesulfonamide

  • MDL number MFCD02625747
  • Molecular Weight : 314.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart