PRIH93ED8507 Princeton BioMolecular Research, Inc.

(E)-(6,7-dimethoxyisoquinolin-1-yl)(3,4-dimethoxyphenyl)methanone oxime

  • MDL number MFCD02625766
  • Molecular Weight : 368.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart