PRIH93ED8519 Princeton BioMolecular Research, Inc.

2-(morpholin-4-ylmethyl)-4-phenyl-5-pyridin-4-yl-2,4-dihydro-3H-1,2,4-triazole-3-thione

  • MDL number MFCD02853077
  • Molecular Weight : 353.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart