PRIH93ED851D Princeton BioMolecular Research, Inc.

methyl 2-({[(1-phenyl-1H-tetrazol-5-yl)thio]acetyl}amino)benzoate

  • MDL number MFCD02689832
  • Molecular Weight : 369.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart