PRIH93ED8526 Princeton BioMolecular Research, Inc.

N-[2,6-dimethyl-3-(morpholin-4-ylsulfonyl)phenyl]methanesulfonamide

  • MDL number MFCD02933858
  • Molecular Weight : 348.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart