PRIH93ED8542 Princeton BioMolecular Research, Inc.

2-[1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]-N-(2-methoxyethyl)acetamide

  • MDL number MFCD03164649
  • Molecular Weight : 414.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart