PRIH93ED854F Princeton BioMolecular Research, Inc.

2-[(2-fluorobenzyl)thio]-3-phenyl-3,5,6,7-tetrahydro-4H-cyclopenta[4,5]thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD02329837
  • Molecular Weight : 408.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart