PRIH93ED8550 Princeton BioMolecular Research, Inc.

2-[(2,6-dichlorobenzyl)thio]-N-phenylacetamide

  • MDL number MFCD03175114
  • Molecular Weight : 326.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart