PRIH93ED8557 Princeton BioMolecular Research, Inc.

N-[2-(4-tert-butylphenoxy)ethyl]-2-nitrobenzamide

  • MDL number MFCD02916540
  • Molecular Weight : 342.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart