PRIH93ED856B Princeton BioMolecular Research, Inc.

2-(2,3-dichlorophenyl)-3-(2-methoxyphenyl)-2,3-dihydroquinazolin-4(1H)-one

  • MDL number MFCD03036790
  • Molecular Weight : 399.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart