PRIH93ED8573 Princeton BioMolecular Research, Inc.

2-({2-[(diphenylmethyl)amino]-2-oxoethyl}thio)-N-1-naphthylacetamide

  • MDL number MFCD00512241
  • Molecular Weight : 440.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart