PRIH93ED8592 Princeton BioMolecular Research, Inc.

N'-((1E)-{4-[methyl(phenyl)amino]phenyl}methylene)-2-(phenylthio)acetohydrazide

  • MDL number MFCD00361444
  • Molecular Weight : 375.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart