PRIH93ED859A Princeton BioMolecular Research, Inc.

N'-[(1Z)-(2-methoxy-1-naphthyl)methylene]benzenesulfonohydrazide

  • MDL number MFCD00384541
  • Molecular Weight : 340.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart