PRIH93ED859B Princeton BioMolecular Research, Inc.

1-(2-oxo-2-phenylethyl)pyridinium

  • MDL number MFCD00459319
  • Molecular Weight : 198.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart