PRIH93ED859C Princeton BioMolecular Research, Inc.

N-(2,6-dimethylphenyl)-N'-phenylthiourea

  • MDL number MFCD00091252
  • Molecular Weight : 256.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart