PRIH93ED85A5 Princeton BioMolecular Research, Inc.

2,2'-(2,3-dihydroxy-1,4-dioxobutane-1,4-diyl)bis(N-methylhydrazinecarbothioamide)

  • MDL number MFCD02063330
  • Molecular Weight : 324.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart