PRIH93ED85A7 Princeton BioMolecular Research, Inc.

N-{4-[(dicyclohexylamino)sulfonyl]phenyl}acetamide

  • MDL number MFCD00495904
  • Molecular Weight : 378.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart