PRIH93ED85B8 Princeton BioMolecular Research, Inc.

1,4-bis[2-(benzyloxy)ethyl]piperazine

  • MDL number MFCD02667717
  • Molecular Weight : 354.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart