PRIH93ED85BA Princeton BioMolecular Research, Inc.

N-cyclohexyl-N'-[3-(2-pyridin-2-ylethyl)phenyl]thiourea

  • MDL number MFCD02575704
  • Molecular Weight : 339.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart