PRIH93ED85D1 Princeton BioMolecular Research, Inc.

N,N'-bis(2-methylphenyl)ethanediamide

  • MDL number MFCD00027571
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart