PRIH93ED85DF Princeton BioMolecular Research, Inc.

N-{2-[(6,7-dimethoxyisoquinolin-1-yl)methyl]-4,5-dimethoxyphenyl}-9H-fluorene-2-sulfonamide

  • MDL number MFCD03267645
  • Molecular Weight : 582.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart