PRIH93ED85E2 Princeton BioMolecular Research, Inc.

5-phenyl-3-(piperidin-1-ylmethyl)-1,3,4-oxadiazole-2(3H)-thione

  • MDL number MFCD00738876
  • Molecular Weight : 275.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart