PRIH93ED85E3 Princeton BioMolecular Research, Inc.

3-(morpholin-4-ylmethyl)-5-phenyl-1,3,4-oxadiazole-2(3H)-thione

  • MDL number MFCD00490492
  • Molecular Weight : 277.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart