PRIH93ED85E4 Princeton BioMolecular Research, Inc.

1,2-dimethoxy-4-[(1Z)-2-nitroprop-1-en-1-yl]benzene

  • MDL number MFCD00138059
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart