PRIH93ED860F Princeton BioMolecular Research, Inc.

2-{2-[(2-methoxy-5-nitrophenyl)amino]-2-oxoethoxy}-N-(4-methoxyphenyl)benzamide

  • MDL number MFCD02333885
  • Molecular Weight : 451.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart