PRIH93ED8617 Princeton BioMolecular Research, Inc.

N-(3-isopropoxypropyl)-N'-1-naphthylurea

  • MDL number MFCD01025428
  • Molecular Weight : 286.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart