PRIH93ED8619 Princeton BioMolecular Research, Inc.

N-(3-chlorophenyl)-N'-[2-(3-chlorophenyl)ethyl]ethanediamide

  • MDL number MFCD07626344
  • Molecular Weight : 337.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart