PRIH93ED8621 Princeton BioMolecular Research, Inc.

N-(1,3-benzodioxol-5-ylmethyl)-4-iodobenzamide

  • MDL number MFCD00422591
  • Molecular Weight : 381.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart