PRIH93ED8632 Princeton BioMolecular Research, Inc.

(4-allyl-5-{[(5-phenyl-1,3,4-oxadiazol-2-yl)methyl]thio}-4H-1,2,4-triazol-3-yl)(phenyl)methanol

  • MDL number MFCD05994255
  • Molecular Weight : 405.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart