PRIH93ED8637 Princeton BioMolecular Research, Inc.

6-butyryl-1,3,3-trimethyl-6-azabicyclo[3.2.1]octane

  • MDL number MFCD00859715
  • Molecular Weight : 223.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart