PRIH93ED8639 Princeton BioMolecular Research, Inc.

6-hexanoyl-1,3,3-trimethyl-6-azabicyclo[3.2.1]octane

  • MDL number MFCD00859737
  • Molecular Weight : 251.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart