PRIH93ED8645 Princeton BioMolecular Research, Inc.

2-[2-isopropyl-5-methyl-4-(1H-tetrazol-1-yl)phenoxy]-N-[4-(2-oxopyrrolidin-1-yl)benzyl]acetamide

  • MDL number MFCD08582793
  • Molecular Weight : 448.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart