PRIH93ED8647 Princeton BioMolecular Research, Inc.

1-chloro-5-isopropyl-4-methoxy-2-methylbenzene

  • MDL number MFCD00027735
  • Molecular Weight : 198.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart