PRIH93ED864A Princeton BioMolecular Research, Inc.

2-oxo-2-(2-thienyl)ethyl thiocyanate

  • MDL number MFCD02063028
  • Molecular Weight : 183.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart