PRIH93ED8664 Princeton BioMolecular Research, Inc.

ethyl 4-{[5-(3-methoxyphenyl)-1-phenyl-1H-1,2,4-triazol-3-yl]carbonyl}piperazine-1-carboxylate

  • MDL number MFCD07606619
  • Molecular Weight : 435.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart