PRIH93ED8671 Princeton BioMolecular Research, Inc.

1,1'-[sulfonylbis(4,1-phenylene)]bis-1H-tetrazole

  • MDL number MFCD01871263
  • Molecular Weight : 354.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart