PRIH93ED8681 Princeton BioMolecular Research, Inc.

1-(diphenylacetyl)-4-(2-furoyl)piperazine

  • MDL number MFCD03274636
  • Molecular Weight : 374.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart