PRIH93ED8688 Princeton BioMolecular Research, Inc.

2-methyl-N'-[(3E)-1-(3-nitrobenzyl)-2-oxo-1,2-dihydro-3H-indol-3-ylidene]-3-furohydrazide

  • MDL number MFCD12130956
  • Molecular Weight : 404.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart