PRIH93ED868F Princeton BioMolecular Research, Inc.

1,3-bis(4-nitrobenzyl)-1H-imidazol-3-ium

  • MDL number MFCD10039602
  • Molecular Weight : 339.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart