PRIH93ED86A7 Princeton BioMolecular Research, Inc.

N,N'-(6-phenylphenanthridine-3,8-diyl)bis(4-butoxybenzamide)

  • MDL number MFCD07065513
  • Molecular Weight : 637.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart