PRIH93ED86AB Princeton BioMolecular Research, Inc.

(1E)-1-[4-(1H-pyrrol-1-yl)phenyl]ethanone O-(2-fluorobenzoyl)oxime

  • MDL number MFCD07065531
  • Molecular Weight : 322.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart