PRIH93ED86B2 Princeton BioMolecular Research, Inc.

2-{[(1-benzyl-1H-benzimidazol-2-yl)thio]methyl}-3-(4-chlorophenyl)quinazolin-4(3H)-one

  • MDL number MFCD09417057
  • Molecular Weight : 509.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart